2-Iodoanisole 95% stabilized

Supplier: Ambeed
Warning

Synonyms: 1-Iodo-2-methoxybenzene

Stabilization: Stabilized with Copper chip

A134637-10G A134637-500G A134637-25G A134637-5G
77109-448EA 9.55 USD
77109-448 77109-450 77314-802 77314-804
2-Iodoanisole 95% stabilized
2-Iodoanisole

Precaución: for Research Use Only

Formula: C₇H₇IO
MW: 234.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001039
CAS Number: 529-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR