N-Hydroxysuccinimide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

A171441-10G A171441-1G A171441-500G A171441-1000G A171441-25G
103809-042EA 62 USD
103809-042 77111-036 77111-038 77111-040 77111-042 77301-128 77301-130
N-Hydroxysuccinimide 98%
N-Hydroxysuccinimide

Precaución: for Research Use Only

Formula: C₄H₅NO₃
MW: 115.09 g/mol
Boiling Pt: 262.4 °C (760 mm Hg)
Melting Pt: 95…98 °C
Flash Pt: 112.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005516
CAS Number: 6066-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR