1-Hydroxypyrene 97%

Supplier: Ambeed
Warning

A360635-1G A360635-25G A360635-5G A360635-100G A360635-10G
103826-264EA 10 USD
103826-264 103837-274 76840-482 76840-484 76840-486 77278-300 77278-302
1-Hydroxypyrene 97%
1-Hydroxypyrene

Precaución: for Research Use Only

Formula: C₁₆H₁₀O
Boiling Pt: 437.4 °C (760 mm Hg)
Flash Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00044543
CAS Number: 5315-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR