1-Hydroxymethyl-3-cyclopentene 97%

Supplier: Ambeed
Warning

A718627-1G A718627-25G A718627-10G A718627-250MG
103840-722EA 31 USD
103840-722 76844-938 76844-940 77243-628 77243-630
1-Hydroxymethyl-3-cyclopentene 97%
1-Hydroxymethyl-3-cyclopentene

Precaución: for Research Use Only

Formula: C₆H₁₀O
Storage Temperature: Refrigerator
MDL Number: MFCD04038630
CAS Number: 25125-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR