1-Isoquinolinol 98%

Supplier: Ambeed
Warning

A297961-100G A297961-10G A297961-250MG A297961-1G A297961-5G A297961-500G
103814-164EA 6 USD
103814-164 103835-308 76693-356 76693-358 76693-360 76837-994 76837-996 77626-526
1-Isoquinolinol 98%
1-Isoquinolinol

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 212…215 °C
Flash Pt: 218.788 °C
Storage Temperature: Ambient
MDL Number: MFCD00006899
CAS Number: 491-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR