1-Hydroxybutanone 96%

Supplier: Ambeed
Danger

Synonyms: 1-Hydroxy-2-butanone, 1-Hydroxybutan-2-one

A214074-1G A214074-5G
103818-366EA 39 USD
103818-366 103849-074 77087-570 77087-572
1-Hydroxybutanone 96%
1-Hydroxybutanone

Precaución: for Research Use Only

Formula: C₄H₈O₂
MW: 88.11 g/mol
Flash Pt: 60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010259
CAS Number: 5077-67-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR