3,5-Dimethyl-1-adamantanol 97%

Supplier: Ambeed
Warning

A546692-1G A546692-25G A546692-5G A546692-100G
103828-662EA 7 USD
103828-662 103839-786 76843-852 77279-962 77307-788 77617-330
3,5-Dimethyl-1-adamantanol 97%
3,5-Dimethyl-1-adamantanol

Precaución: for Research Use Only

Formula: C₁₂H₂₀O
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00074775
CAS Number: 707-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR