1-Hydroxy-2-naphthoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Hydroxy-naphthalene-2-carboxylic acid
A155867-100G
A155867-500G
A155867-25G
A155867-1000G
103808-826EA
89
USD
103808-826
76871-856
76871-858
77300-150
77318-750
1-Hydroxy-2-naphthoic acid 98%
1-Hydroxy-2-naphthoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₈O₃ Melting Pt: 195…200 °C Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003960 CAS Number: 86-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...