1-Hydroxy-1-cyclopropanecarboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-hydroxycyclopropane-1-carboxylic acid, 1-Hydroxycyclopropanecarboxylic acid
A135520-100G
A135520-100MG
A135520-10G
A135520-250MG
A135520-1G
A135520-25G
A135520-5G
103802-170EA
15
USD
103802-170
76870-956
76870-958
76870-960
76870-962
77315-002
77315-004
77315-006
1-Hydroxy-1-cyclopropanecarboxylic acid 95%
1-Hydroxy-1-cyclopropanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₄H₆O₃ MW: 102.089 g/mol Boiling Pt: 276.6 °C (760 mm Hg) Melting Pt: 108…110 °C Flash Pt: 135.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010797 CAS Number: 17994-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...