1H-Pyrrolo[3,2-d]pyrimidin-4(5H)-one 98%

Supplier: Ambeed
Warning

A213561-100MG A213561-10G A213561-250MG A213561-1G A213561-25G A213561-5G
103818-308EA 66 USD
103818-308 103849-016 77192-704 77192-706 77192-708 77303-298 77303-300 77303-302
1H-Pyrrolo[3,2-d]pyrimidin-4(5H)-one 98%
1H-Pyrrolo[3,2-d]pyrimidin-4(5H)-one

Precaución: for Research Use Only

Formula: C₆H₅N₃O
Boiling Pt: 392.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09037890
CAS Number: 5655-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR