1H-Pyrrolo[3,2-b]pyridine-7-carboxylic acid 97%

Supplier: Ambeed
Warning

A485997-1G A485997-100MG A485997-250MG
103828-176EA 357 USD
103828-176 103839-250 76843-190 76991-044 76991-046
1H-Pyrrolo[3,2-b]pyridine-7-carboxylic acid 97%
1H-Pyrrolo[3,2-b]pyridine-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Boiling Pt: 399.1±22.0 °C (760 mm Hg)
Flash Pt: 195.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12963157
CAS Number: 1190319-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR