1H-Pyrrolo[3,2-b]pyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Azaindole-3-carboxaldehyde

A178433-10G A178433-250MG A178433-1G A178433-25G A178433-5G
103807-330EA 31 USD
103807-330 103845-670 77029-334 77029-336 77083-096 77083-098 77083-100
1H-Pyrrolo[3,2-b]pyridine-3-carbaldehyde 97%
1H-Pyrrolo[3,2-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
Boiling Pt: 361.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11877845
CAS Number: 276862-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR