4-Azaindole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrolo[3,2-b]pyridine-2-carboxylic acid, 2-Carboxy-1H-pyrrolo[3,2b]pyridine

A144994-100MG A144994-250MG A144994-5G
103803-476EA 34 USD
103803-476 103841-554 76811-932 76811-934 77316-820
4-Azaindole-2-carboxylic acid 98%
4-Azaindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.145 g/mol
Boiling Pt: 446.8 °C (760 mm Hg)
Flash Pt: 224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09955613
CAS Number: 17288-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR