1H-Pyrrolo[3,2-b]pyridin-5-amine hydrochloride 97%

Supplier: Ambeed
Warning

A109720-100MG A109720-250MG A109720-1G
103797-552EA 164 USD
103797-552 103823-358 76821-738 76821-740 77310-166
1H-Pyrrolo[3,2-b]pyridin-5-amine hydrochloride 97%
1H-Pyrrolo[3,2-b]pyridin-5-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClN₃
MW: 169.6100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16875696
CAS Number: 1260771-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR