1H-Pyrrolo[2,3-c]pyridine-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A629845-100MG A629845-250MG
103840-270EA 698 USD
103840-270 76831-160 77684-515
1H-Pyrrolo[2,3-c]pyridine-5-carbaldehyde 97%
1H-Pyrrolo[2,3-c]pyridine-5-carbaldehyde

Precaución: For research use only.

Formula: C₈H₆N₂O
MW: 146.146 g/mol
Boiling Pt: 353.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10697530
CAS Number: 130473-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR