1H-Pyrrolo[2,3-c]pyridine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A206946-100MG A206946-250MG
103817-746EA 516 USD
103817-746 77097-568 77097-570
1H-Pyrrolo[2,3-c]pyridine-4-carboxylic acid 95%
1H-Pyrrolo[2,3-c]pyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12963371
CAS Number: 1190319-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR