1H-Pyrrolo[2,3-c]pyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A224327-10G A224327-1G A224327-5G A224327-250MG
103819-100EA 31 USD
103819-100 103849-990 76814-992 77088-638 77088-640 77883-578
1H-Pyrrolo[2,3-c]pyridine-3-carbaldehyde 97%
1H-Pyrrolo[2,3-c]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
Boiling Pt: 383.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08272238
CAS Number: 25957-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR