6-Azaindole 98%

Supplier: Ambeed
Warning

A216265-1G A216265-25G A216265-5G A216265-10G A216265-250MG
103818-548EA 8 USD
103818-548 103849-264 77087-838 77087-840 77087-842 77139-756 77139-758
6-Azaindole 98%
6-Azaindole

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 295.7 °C (760 mm Hg)
Melting Pt: 136…137 °C
Flash Pt: 137.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01686899
CAS Number: 271-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR