1H-Pyrrolo[2,3-c]pyridin-5-amine 98%

Supplier: Ambeed
Warning

A210245-100MG A210245-250MG A210245-50MG
103817-990EA 84 USD
103817-990 103848-684 76785-662 76785-664 76785-666
1H-Pyrrolo[2,3-c]pyridin-5-amine 98%
1H-Pyrrolo[2,3-c]pyridin-5-amine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 388.5 °C (760 mm Hg)
Flash Pt: 217.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00817668
CAS Number: 174610-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR