1H-Pyrrolo[2,3-b]pyridine-5-carbaldehyde 98%

Supplier: Ambeed
Warning

A172730-1G A172730-5G A172730-100MG A172730-250MG
103806-740EA 65 USD
103806-740 103845-034 77082-244 77082-246 77163-618 77163-620
1H-Pyrrolo[2,3-b]pyridine-5-carbaldehyde 98%
1H-Pyrrolo[2,3-b]pyridine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD08272241
CAS Number: 849067-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR