1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Azaindole-3-carboxylic acid
A163883-10G
A163883-250MG
A163883-1G
A163883-25G
A163883-5G
A163883-100G
103805-780EA
6
USD
103805-780
103844-044
77235-996
77235-998
77320-164
77320-166
77320-168
77615-346
1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid 97%
1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 446.8 °C (760 mm Hg) Melting Pt: 205…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD07778360 CAS Number: 156270-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...