1H-Pyrrolo[2,3-b]pyridine-3-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 7-Azaindole-3-carboxaldehyde

A266183-10G A266183-25G A266183-5G A266183-1G
103812-788EA 6 USD
103812-788 103852-688 76772-012 77092-390 77092-392 77304-950
1H-Pyrrolo[2,3-b]pyridine-3-carbaldehyde 98%
1H-Pyrrolo[2,3-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.1460 g/mol
Boiling Pt: 361.1 °C (760 mm Hg)
Melting Pt: 216…220 °C at N/A mmHg
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD03407363
CAS Number: 4649-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR