1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A101771-100MG
A101771-10G
A101771-250MG
A101771-25G
A101771-1G
A101771-5G
103796-128EA
53
USD
103796-128
103821-816
77233-090
77233-092
77233-094
77233-096
77282-826
77282-828
1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid 97%
1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ Boiling Pt: 446.8 °C (760 mm Hg) Melting Pt: 285 °C Flash Pt: 224.03 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659752 CAS Number: 136818-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...