1H-Pyrrolo[2,3-b]pyridine-2,3-dione 96%

Supplier: Ambeed
Warning

A151645-100MG A151645-250MG A151645-5G A151645-1G
103804-290EA 224 USD
103804-290 77235-454 77235-456 77235-458 77318-028
1H-Pyrrolo[2,3-b]pyridine-2,3-dione 96%
1H-Pyrrolo[2,3-b]pyridine-2,3-dione

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂
MW: 148.119 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09999197
CAS Number: 5654-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR