7-Azaindole 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

A156097-100G A156097-10G A156097-500G A156097-25G A156097-5G
103843-032EA 7 USD
103843-032 76768-332 76768-334 76768-336 77318-772 77318-774
7-Azaindole 98%
7-Azaindole

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 273.8 °C (760 mm Hg)
Melting Pt: 105…107 °C
Flash Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00005606
CAS Number: 271-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR