1H-Pyrrolo[2,3-b]pyridin-6-amine 97%

Supplier: Ambeed
Warning

A105252-250MG A105252-5G
103796-864EA 45 USD
103796-864 103822-592 76809-548 77309-462
1H-Pyrrolo[2,3-b]pyridin-6-amine 97%
1H-Pyrrolo[2,3-b]pyridin-6-amine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.1506 g/mol
Melting Pt: 126.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272224
CAS Number: 145901-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR