1H-Pyrrolo[2,3-b]pyridin-5-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-7-azaindole

A411096-1G A411096-25G A411096-5G A411096-100G A411096-10G A411096-250MG A411096-500G
103827-220EA 6 USD
103827-220 103838-270 76841-800 76841-802 76841-804 77155-712 77155-714 77155-716 77155-718
1H-Pyrrolo[2,3-b]pyridin-5-ol 98%
1H-Pyrrolo[2,3-b]pyridin-5-ol

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272242
CAS Number: 98549-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR