1H-Pyrrole-3-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Pyrrole-3-carboxaldehyde

A476821-1G A476821-5G A476821-10G A476821-250MG A476821-100G
103828-086EA 8 USD
103828-086 103839-156 76843-084 76843-086 77617-066 77617-068 77746-980
1H-Pyrrole-3-carbaldehyde 97%
1H-Pyrrole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅NO
MW: 95.10 g/mol
Melting Pt: 65…68 °C
Flash Pt: 103.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426733
CAS Number: 7126-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR