2-Pyrrolecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrole-2-carboxaldehyde, Pyrrole-2-carboxaldehyde, 1H-Pyrrole-2-carbaldehyde, Pyrrole-2-carbaldehyde

A509828-25G A509828-5G A509828-100G A509828-10G A509828-500G
103839-478EA 6 USD
103839-478 76843-466 76843-468 77017-288 77017-290 77017-292
2-Pyrrolecarbaldehyde 98%
2-Pyrrolecarbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 217…219 °C (760 mm Hg)
Melting Pt: 43…46 °C
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005217
CAS Number: 1003-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR