1H-Pyrrole-2,5-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A163689-100MG A163689-250MG A163689-25G A163689-50MG A163689-1G A163689-5G
103805-746EA 69 USD
103805-746 103844-008 76985-162 76985-164 76985-166 76985-168 77320-114 77320-116
1H-Pyrrole-2,5-dicarboxylic acid 97%
1H-Pyrrole-2,5-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₄
Boiling Pt: 528.2±35.0 °C (760 mm Hg)
Melting Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09991932
CAS Number: 937-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR