1H-Pyrazolo[4,3-b]pyridine-5-carbonitrile 98%

Supplier: Ambeed
Warning

A249058-100MG A249058-250MG
103811-852EA 269 USD
103811-852 103851-688 76815-612 76815-614
1H-Pyrazolo[4,3-b]pyridine-5-carbonitrile 98%
1H-Pyrazolo[4,3-b]pyridine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₄
MW: 144.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384475
CAS Number: 1033772-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR