1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A162465-100MG A162465-250MG A162465-1G A162465-5G
103805-618EA 98 USD
103805-618 103843-868 76973-636 76973-638 77300-592 77300-594
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid 97%
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193615
CAS Number: 1260648-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR