1H-Pyrazolo[3,4-d]pyrimidin-4-amine 98%

Supplier: Ambeed
Danger

A159710-10G A159710-1G A159710-25G A159710-5G
103843-502EA 6 USD
103843-502 77011-164 77011-166 77319-402 77319-404
1H-Pyrazolo[3,4-d]pyrimidin-4-amine 98%
1H-Pyrazolo[3,4-d]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₅N₅
Boiling Pt: 299.5 °C (760 mm Hg)
Melting Pt: 325 °C at N/A mmHg
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005688
CAS Number: 2380-63-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR