1H-Pyrazolo[3,4-c]pyridine 98%

Supplier: Ambeed
Warning

A217118-250MG A217118-1G
103818-602EA 14 USD
103818-602 103849-330 76986-924 77087-946
1H-Pyrazolo[3,4-c]pyridine 98%
1H-Pyrazolo[3,4-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.1240 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00661308
CAS Number: 271-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR