1H-Pyrazolo[3,4-b]pyridine-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A159774-100MG
A159774-250MG
A159774-5G
103805-282EA
45
USD
103805-282
76783-784
77871-799
77871-800
1H-Pyrazolo[3,4-b]pyridine-3-carboxylic acid 97%
1H-Pyrazolo[3,4-b]pyridine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₅N₃O₂ Boiling Pt: 475 °C (760 mm Hg) Melting Pt: 265…270 °C Flash Pt: 241.107 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11040725 CAS Number: 116855-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...