1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 96%

Supplier: Ambeed
Warning

A218737-100MG A218737-250MG A218737-1G A218737-5G
103818-738EA 70 USD
103818-738 103849-608 76814-826 76814-828 77088-134 77088-136
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 96%
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₄
Storage Temperature: Refrigerator
MDL Number: MFCD13176785
CAS Number: 956010-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR