1H-Pyrazolo[3,4-b]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 7-Azaindazole, Pyrazolo[3,4-b]pyridine

A131193-100G A131193-10G A131193-1G A131193-25G A131193-5G
103801-538EA 9 USD
103801-538 77161-884 77161-886 77298-458 77298-460 77298-462
1H-Pyrazolo[3,4-b]pyridine 97%
1H-Pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.12 g/mol
Boiling Pt: 294.5 °C (760 mm Hg)
Melting Pt: 99…101 °C
Flash Pt: 145.716 °C
Storage Temperature: Ambient
MDL Number: MFCD05663981
CAS Number: 271-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR