1H-Pyrazolo[3,4-b]pyridin-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 1H-pyrazolo[3,4-b]pyridin-3-amine, 3-Amino-1H-pyrazolo[3,4-b]pyridine

A161129-100G A161129-10G A161129-250MG A161129-1G A161129-25G A161129-5G
103805-454EA 19 USD
103805-454 103843-692 77163-182 77163-184 77163-186 77319-652 77319-654 77319-656
1H-Pyrazolo[3,4-b]pyridin-3-amine 95%
1H-Pyrazolo[3,4-b]pyridin-3-amine

Precaución: Keep in a dark place, inert atmosphere, room temperature

Formula: C₆H₆N₄
MW: 134.14 g/mol
Boiling Pt: 406.7 °C (760 mm Hg)
Melting Pt: 187…192 °C
Flash Pt: 228.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04113611
CAS Number: 6752-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR