1H-Pyrazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A118060-100G A118060-10G A118060-25G A118060-500G A118060-1G A118060-5G
103798-980EA 6 USD
103798-980 103824-874 77094-314 77094-316 77094-318 77094-320 77311-662 77311-664
1H-Pyrazole-4-carboxylic acid 98%
1H-Pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂
Boiling Pt: 417.1 °C (760 mm Hg)
Melting Pt: 282 °C
Flash Pt: 206.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00011558
CAS Number: 37718-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR