1H-Pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A435381-1G A435381-25G A435381-5G A435381-10G A435381-250MG
103827-596EA 15 USD
103827-596 103838-664 76842-384 76842-386 76842-388 77168-720 77168-722
1H-Pyrazole-4-carbaldehyde 97%
1H-Pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 96.0900 g/mol
Boiling Pt: 300 °C (760 mm Hg)
Melting Pt: 150…154 °C
Flash Pt: 139.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179514
CAS Number: 35344-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR