1H-Pyrazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Pyrazolecarboxylic acid, Pyrazole-3-carboxylic acid

A116703-100G A116703-10G A116703-25G A116703-500G A116703-5G
103824-652EA 8 USD
103824-652 77094-246 77094-248 77094-250 77094-252 77311-446
1H-Pyrazole-3-carboxylic acid 98%
1H-Pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Boiling Pt: 417.1 °C (760 mm Hg)
Melting Pt: 211 °C
Flash Pt: 206.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00077436
CAS Number: 1621-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR