3,5-Pyrazoledicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Pyrazole-3,5-dicarboxylic acid, 1H-Pyrazole-3,5-dicarboxylic acid

A100909-100G A100909-10G A100909-250MG A100909-25G A100909-1G
103795-688EA 6 USD
103795-688 103821-378 77008-288 77008-290 77008-292 77008-294 77296-904
3,5-Pyrazoledicarboxylic acid 97%
3,5-Pyrazoledicarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
Boiling Pt: 390 °C (760 mm Hg)
Flash Pt: 325.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005235
CAS Number: 3112-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR