1H-Pyrazol-4-ol 97%

Supplier: Ambeed
Warning

A120629-100MG A120629-250MG A120629-1G A120629-5G A120629-10G
103799-972EA 49 USD
103799-972 76870-206 76870-208 77312-078 77312-080 77566-236
1H-Pyrazol-4-ol 97%
1H-Pyrazol-4-ol

Precaución: For research use only.

Formula: C₃H₄N₂O
MW: 84.08 g/mol
Boiling Pt: 326.1 °C (760 mm Hg)
Flash Pt: 151 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01693118
CAS Number: 4843-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR