1H-Indole-7-carbonitrile 97%

Supplier: Ambeed
Warning

A378351-10G A378351-250MG A378351-1G A378351-25G A378351-5G
103826-616EA 15 USD
103826-616 103837-650 76828-982 76828-984 76840-964 76840-966 76840-968
1H-Indole-7-carbonitrile 97%
1H-Indole-7-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂
MW: 142.1573 g/mol
Melting Pt: 95…99 °C
Flash Pt: 121.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00800653
CAS Number: 96631-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR