Indole-7-carbaldehyd 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Indole-7-carbaldehyde, Indole-7-carboxaldehyde

A701082-1G A701082-25G A701082-5G A701082-100G A701082-10G A701082-250MG
103840-642EA 20 USD
103840-642 76844-858 76844-860 76844-862 77267-592 77267-594 77267-596
Indole-7-carbaldehyd 98%
Indole-7-carbaldehyd

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 87…91 °C
Flash Pt: 166.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01318152
CAS Number: 1074-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR