Indole-6-carbaldehyd 97%

Supplier: Ambeed
Warning

Synonyms: Indole-6-carboxaldehyde, 1H-Indole-6-carbaldehyde

A146949-10G A146949-1G A146949-5G
103803-724EA 9 USD
103803-724 103841-820 77259-068 77317-176 77317-178
Indole-6-carbaldehyd 97%
Indole-6-carbaldehyd

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 127…131 °C
Flash Pt: 166.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02179596
CAS Number: 1196-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR