Indole-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-Indole-5-carboxylic acid
A394775-1G
A394775-5G
A394775-250MG
A394775-25G
A394775-10G
103826-948EA
6
USD
103826-948
103837-986
76841-406
76841-408
77278-682
77307-032
77572-204
Indole-5-carboxylic acid 98%
Indole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₇NO₂ MW: 161.16 g/mol Melting Pt: 211…213 °C Flash Pt: 207.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005678 CAS Number: 1670-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...