Indole-3-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Indole-3-carbonitrile, 3-Cyano-1H-indole, 3-Cyanoindole

A150460-100G A150460-10G A150460-25G A150460-5G
103842-304EA 8 USD
103842-304 77259-228 77259-230 77299-806 77317-822
Indole-3-carbonitrile 97%
Indole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂
MW: 142.16 g/mol
Boiling Pt: 350 °C (760 mm Hg)
Melting Pt: 179…182 °C
Flash Pt: 121.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00022717
CAS Number: 5457-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR