1H-Indole-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: Indole-2-carbonitrile

A175090-1G A175090-5G
103806-998EA 23 USD
103806-998 103845-302 77082-622 77082-624
1H-Indole-2-carbonitrile 97%
1H-Indole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂
MW: 142.161 g/mol
Boiling Pt: 350.0±15.0 °C (760 mm Hg)
Melting Pt: 101 °C
Flash Pt: 121.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00187566
CAS Number: 36193-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR