Indole-2-carbaldehyd 97%

Supplier: Ambeed
Warning

Synonyms: Indole-2-carboxaldehyde, 1H-Indole-2-carbaldehyde

A147460-100MG A147460-10G A147460-250MG A147460-1G A147460-25G A147460-5G
103803-780EA 15 USD
103803-780 103841-882 77010-606 77010-608 77010-610 77299-580 77317-256 77317-258
Indole-2-carbaldehyd 97%
Indole-2-carbaldehyd

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 339.1 °C (760 mm Hg)
Melting Pt: 138…142 °C
Flash Pt: 166.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03001425
CAS Number: 19005-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR